2-Acrylamido-2-methylpropanesulfonic acid, 97%
2-Acrylamido-2-methylpropanesulfonic acid, 97%
2-Acrylamido-2-methylpropanesulfonic acid, 97%
Thermo Scientific Chemicals

2-Acrylamido-2-methylpropanesulfonic acid, 97%

CAS: 15214-89-8 | C7H13NO4S | 207.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
40029050050 gGlass bottle
Catalog number 400290500
Price (EUR)
20,80
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
20,80
Each
Chemical Identifiers
CAS15214-89-8
IUPAC Name2-methyl-2-(prop-2-enamido)propane-1-sulfonic acid
Molecular FormulaC7H13NO4S
InChI KeyXHZPRMZZQOIPDS-UHFFFAOYSA-N
SMILESCC(C)(CS(O)(=O)=O)NC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or chips and/or chunks
Infrared spectrumConforms
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only