2'-Aminoacetophenone, 97%
2'-Aminoacetophenone, 97%
2'-Aminoacetophenone, 97%
Thermo Scientific Chemicals

2'-Aminoacetophenone, 97%

CAS: 551-93-9 | C8H9NO | 135.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 400620500
Price (EUR)
209,00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
209,00
Each
Chemical Identifiers
CAS551-93-9
IUPAC Name1-(2-aminophenyl)ethan-1-one
Molecular FormulaC8H9NO
InChI KeyGTDQGKWDWVUKTI-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.6130 to 1.6170 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only