Anthrone, 95%, pure
Anthrone, 95%, pure
Anthrone, 95%, pure
Thermo Scientific Chemicals

Anthrone, 95%, pure

CAS: 90-44-8 | C14H10O | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 401360250
Price (EUR)
79,65
Precio exclusivo compra web
88,10
Save 8,45 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
79,65
Precio exclusivo compra web
88,10
Save 8,45 (10%)
Each
Chemical Identifiers
CAS90-44-8
IUPAC Name9,10-dihydroanthracen-9-one
Molecular FormulaC14H10O
InChI KeyRJGDLRCDCYRQOQ-UHFFFAOYSA-N
SMILESO=C1C2=CC=CC=C2CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Melting point155°C to 161°C
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only