Bis(2-ethylhexyl) adipate, 99%
Bis(2-ethylhexyl) adipate, 99%
Bis(2-ethylhexyl) adipate, 99%
Thermo Scientific Chemicals

Bis(2-ethylhexyl) adipate, 99%

CAS: 103-23-1 | C22H42O4 | 370.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 402462500
Price (EUR)
21,60
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
21,60
Each
Chemical Identifiers
CAS103-23-1
IUPAC Name1,6-bis(2-ethylhexyl) hexanedioate
Molecular FormulaC22H42O4
InChI KeySAOKZLXYCUGLFA-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<30 APHA
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4440 to 1.4500 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only