Bis(2-ethylhexyl) hydrogen phosphate, 95%
Bis(2-ethylhexyl) hydrogen phosphate, 95%
Bis(2-ethylhexyl) hydrogen phosphate, 95%
Thermo Scientific Chemicals

Bis(2-ethylhexyl) hydrogen phosphate, 95%

CAS: 298-07-7 | C16H35O4P | 322.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 402470050
Price (EUR)
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS298-07-7
IUPAC Namebis[(2-ethylhexyl)oxy]phosphinic acid
Molecular FormulaC16H35O4P
InChI KeySEGLCEQVOFDUPX-UHFFFAOYNA-N
SMILESCCCCC(CC)COP(O)(=O)OCC(CC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4400 to 1.4440 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Titration with NaOH>=94 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only