tert-Butyl isocyanide, 97%
tert-Butyl isocyanide, 97%
tert-Butyl isocyanide, 97%
Thermo Scientific Chemicals

tert-Butyl isocyanide, 97%

CAS: 7188-38-7 | C5H9N | 83.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5g
1 g
Packaging:
Glass bottle
Catalog number 403600010
Price (EUR)
43,20
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
43,20
Each
Specifications
Assay Percent Range96% min. (GC)
CAS7188-38-7
Chemical Name or Materialtert-Butyl isocyanide
Density0.7300g/mL
EINECS Number230-553-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only