2-Chloronicotinic acid, 99%
2-Chloronicotinic acid, 99%
2-Chloronicotinic acid, 99%
Thermo Scientific Chemicals

2-Chloronicotinic acid, 99%

CAS: 2942-59-8 | C6H4ClNO2 | 157.56 g/mol
Have Questions?
Catalog NumberQuantityPackaging
40486050050 gGlass bottle
Catalog number 404860500
Price (EUR)
82,10
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
82,10
Each
Chemical Identifiers
CAS2942-59-8
IUPAC Name2-chloropyridine-3-carboxylic acid
Molecular FormulaC6H4ClNO2
InChI KeyIBRSSZOHCGUTHI-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CN=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Form)Powder
Titration with NaOH>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only