4-Chlorophenylhydrazine hydrochloride, 97%
4-Chlorophenylhydrazine hydrochloride, 97%
4-Chlorophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals

4-Chlorophenylhydrazine hydrochloride, 97%

CAS: 1073-70-7 | C6H7ClN2·HCl | 179.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 404950050
Price (EUR)
31,10
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
31,10
Each
Chemical Identifiers
CAS1073-70-7
IUPAC Namehydrogen (4-chlorophenyl)hydrazine chloride
Molecular FormulaC6H8Cl2N2
InChI KeyYQVZREHUWCCHHX-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige to pink to light brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Water=<3 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only