Chromium(VI) oxide, 98+%, ACS reagent
Chromium(VI) oxide, 98+%, ACS reagent
Chromium(VI) oxide, 98+%, ACS reagent
Thermo Scientific Chemicals

Chromium(VI) oxide, 98+%, ACS reagent

CAS: 1333-82-0 | CrO3 | 99.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 405235000
Price (EUR)
103,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
103,00
Each
Chemical Identifiers
CAS1333-82-0
IUPAC Nametrioxochromium
Molecular FormulaCrO3
InChI KeyWGLPBDUCMAPZCE-UHFFFAOYSA-N
SMILESO=[Cr](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder, crystals or granules
Insoluble matter=<0.01 %
Titration Iodometric>=98.0 %
Iron (Fe)=<200 ppm
Barium (Ba)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only