Crystal Violet, certified
Crystal Violet, certified
Crystal Violet, certified
Thermo Scientific Chemicals

Crystal Violet, certified

CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 405835000
Price (EUR)
217,65
Online exclusive
242,00
Save 24,35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
217,65
Online exclusive
242,00
Save 24,35 (10%)
Each
Chemical Identifiers
CAS548-62-9, 90-94-8
Specifications
Absorption Ratio0.98 to 1.2 (P-15/P+15)
Beilstein13, 756
CAS548-62-9
CAS90-94-8
ChEBICHEBI:41688
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only