6,9-Diamino-2-ethoxyacridine lactate monohydrate, 95%, Thermo Scientific Chemicals
6,9-Diamino-2-ethoxyacridine lactate monohydrate, 95%, Thermo Scientific Chemicals
6,9-Diamino-2-ethoxyacridine lactate monohydrate, 95%, Thermo Scientific Chemicals
6,9-Diamino-2-ethoxyacridine lactate monohydrate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6,9-Diamino-2-ethoxyacridine lactate monohydrate, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
406381000100 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 406381000
Price (EUR)
287,00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
287,00
Each
Add to cart
Chemical Identifiers
CAS6402-23-9
IUPAC Name2-hydroxypropanoic acid 7-ethoxyacridine-3,9-diamine hydrate
Molecular FormulaC18H23N3O5
InChI KeyNYEPHMYJRNWPLA-UHFFFAOYNA-N
SMILESO.CC(O)C(O)=O.CCOC1=CC=C2N=C3C=C(N)C=CC3=C(N)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO4>=94 %
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only