2,5-Dimethyl-p-benzoquinone, 99%
2,5-Dimethyl-p-benzoquinone, 99%
2,5-Dimethyl-p-benzoquinone, 99%
Thermo Scientific Chemicals

2,5-Dimethyl-p-benzoquinone, 99%

CAS: 137-18-8 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 408230250
Price (EUR)
273,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
273,00
Each
Chemical Identifiers
CAS137-18-8
IUPAC Name2,5-dimethylcyclohexa-2,5-diene-1,4-dione
Molecular FormulaC8H8O2
InChI KeyMYKLQMNSFPAPLZ-UHFFFAOYSA-N
SMILESCC1=CC(=O)C(C)=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point123°C to 126°C
Appearance (Color)Yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only