N,N-Dimethylcyclohexylamine, 99%
N,N-Dimethylcyclohexylamine, 99%
N,N-Dimethylcyclohexylamine, 99%
Thermo Scientific Chemicals

N,N-Dimethylcyclohexylamine, 99%

CAS: 98-94-2 | C8H17N | 127.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 408250025
Price (EUR)
182,00
Each
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
182,00
Each
Chemical Identifiers
CAS98-94-2
IUPAC NameN,N-dimethylcyclohexanamine
Molecular FormulaC8H17N
InChI KeySVYKKECYCPFKGB-UHFFFAOYSA-N
SMILESCN(C)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.4525 to 1.4545 (20°C, 589 nm)
Color scale=<50 APHA
Water=<0.25 %
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only