N,N-Dimethylguanidine sulfate, 98%
N,N-Dimethylguanidine sulfate, 98%
N,N-Dimethylguanidine sulfate, 98%
N,N-Dimethylguanidine sulfate, 98%
Thermo Scientific Chemicals

N,N-Dimethylguanidine sulfate, 98%

CAS: 598-65-2 | 0·5 H2SO4 | 272.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
100 g
Packaging:
Plastic bottle
Catalog number 408350500
Price (EUR)
169,00
Each
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
169,00
Each
Add to cart
Chemical Identifiers
CAS598-65-2
IUPAC NameN,N-dimethylguanidine; sulfuric acid
Molecular FormulaC3H11N3O4S
InChI KeyACPFLXJEPKOPGF-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.CN(C)C(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only