2,4-Dimethylphenol, 99%
2,4-Dimethylphenol, 99%
2,4-Dimethylphenol, 99%
Thermo Scientific Chemicals

2,4-Dimethylphenol, 99%

CAS: 105-67-9 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 408451000
Price (EUR)
65,70
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
65,70
Each
Chemical Identifiers
CAS105-67-9
IUPAC Name2,4-dimethylphenol
Molecular FormulaC8H10O
InChI KeyKUFFULVDNCHOFZ-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5380 to 1.5420 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only