Methyldiphenylphosphine, 99%
Methyldiphenylphosphine, 99%
Methyldiphenylphosphine, 99%
Thermo Scientific Chemicals

Methyldiphenylphosphine, 99%

CAS: 1486-28-8 | C13H13P | 200.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 408970250
Price (EUR)
186,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
186,00
Each
Chemical Identifiers
CAS1486-28-8
IUPAC Namemethyldiphenylphosphane
Molecular FormulaC13H13P
InChI KeyUJNZOIKQAUQOCN-UHFFFAOYSA-N
SMILESCP(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.6230 to 1.6260 (20°C, 589 nm)
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only