Gelatin, for analysis, granular
Thermo Scientific Chemicals

Gelatin, for analysis, granular

Gelatin, CAS # 9000-70-8, also known as gelatin type A, derived from porcine skin, is a gelling compound used in electrophoresis and in vitro cell cultures. | CAS: 9000-70-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 410875000
Price (EUR)
44,65
Online Exclusive
49,10
Save 4,45 (9%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
44,65
Online Exclusive
49,10
Save 4,45 (9%)
Each
Chemical Identifiers
CAS9000-70-8
MDL NumberMFCD00081638
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<2 %
pH3.8 to 7.6 (Ph. Eur.(c=1, about 55°C))
Arsenic (As)=<1 ppm
Lead (Pb)=<5 ppm
Sulfur dioxide (SO2)=<40 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Gelatin derived from porcine skin is a powder created from the acidic digestion of collagen and is also known as gelatin type A

• It comprises heterogeneous mixture of water-soluble proteins of high average molecular masses, including glycine, proline and hydroxyproline

Applications

• Gelatin has been used in in vitro coating cell culture to improve attachment of cell lines • It is added to PCR reactions in order to stabilize DNA Taq polymerase

• It can be used as a blocking reagent in several biological techniques including western blotting, ELISA, and immunochemistry

• It can be applied in microencapsulation assays to delivery vehicle for release of active biomolecules since it is a biocompatible polymer

• It is used as a component of media for species differentiation in bacteriology and in light microscope autoradiography

• It has been used in the generation of scaffolds for tissue engineering assays

RUO – Research Use Only

General References:

  1. Nishi, N.; Inui, M.; Miyanaka, H.; Oya, H., Wada, F. Western blot analysis of epidermal growth factor using gelatin-coated polyvinylidene difluoride membranes. Anal Biochem. 1995, 20, 227 (2), 401-402.
  2. Sarika, P. R.; Pavithran, A.; James, N. R. Cationized gelatin/gum arabic polyelectrolyte complex: Study of electrostatic interactions. Food Hydrocolloids 2015, 49, 176-182.
  3. Chen, P. Y.; Yang, K. C.; Wu, C. C.; Yu, J. H.; Lin, F. H.; Sun, J. S. Fabrication of large perfusable macroporous cell-laden hydrogel scaffolds using microbial transglutaminase. Acta Biomater. 2014, 10 (2), 912-920.