Hexadecyltrimethylammonium Chloride, 97%
Hexadecyltrimethylammonium Chloride, 97%
Hexadecyltrimethylammonium Chloride, 97%
Thermo Scientific Chemicals

Hexadecyltrimethylammonium Chloride, 97%

CAS: 112-02-7 | C19H42ClN | 320.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 411410250
Price (EUR)
102,65
Online Exclusive
114,00
Save 11,35 (10%)
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
102,65
Online Exclusive
114,00
Save 11,35 (10%)
Each
Chemical Identifiers
CAS112-02-7
IUPAC Namehexadecyltrimethylazanium chloride
Molecular FormulaC19H42ClN
InChI KeyWOWHHFRSBJGXCM-UHFFFAOYSA-M
SMILES[Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Argentometric96.0 to 104.0 %
Appearance (Color)White to almost white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only