1,6-Hexanediamine, 60 wt % aqueous soln.
1,6-Hexanediamine, 60 wt % aqueous soln.
1,6-Hexanediamine, 60 wt % aqueous soln.
Thermo Scientific Chemicals

1,6-Hexanediamine, 60 wt % aqueous soln.

CAS: 124-09-4 | C6H16N2 | 116.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 411562500
Price (EUR)
33,80
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
33,80
Each
Chemical Identifiers
CAS124-09-4
IUPAC Namehexane-1,6-diamine
Molecular FormulaC6H16N2
InChI KeyNAQMVNRVTILPCV-UHFFFAOYSA-N
SMILESNCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Strength59 to 61 wt%
Appearance (Form)Clear solution
Color scale=<20 APHA
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only