Hydrazine acetate, 97%
Hydrazine acetate, 97%
Hydrazine acetate, 97%
Thermo Scientific Chemicals

Hydrazine acetate, 97%

CAS: 7335-65-1 | C2H8N2O2 | 92.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 411750050
Price (EUR)
59,65
キャンペーン価格
66,00
Save 6,35 (10%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
59,65
キャンペーン価格
66,00
Save 6,35 (10%)
Each
Chemical Identifiers
CAS7335-65-1
IUPAC Nameacetic acid; hydrazine
Molecular FormulaC2H8N2O2
InChI KeyYFHNDHXQDJQEEE-UHFFFAOYSA-N
SMILESNN.CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Adhering powder or crystals
Infrared spectrumConforms
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only