Hydriodic acid, ACS reagent, 55% in water, without stabilizer
Hydriodic acid, ACS reagent, 55% in water, without stabilizer
Thermo Scientific Chemicals

Hydriodic acid, ACS reagent, 55% in water, without stabilizer

CAS: 10034-85-2 | HI | 127.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 411840500
Price (EUR)
-
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS10034-85-2
IUPAC Namehydrogen iodide
Molecular FormulaHI
InChI KeyXMBWDFGMSWQBCA-UHFFFAOYSA-N
SMILESI
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<10 ppm
Appearance (Color)Dark red-brown
Appearance (Form)Solution
Titration with NaOH55.0 to 58.0 %
Residue after ignition=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only