2-Imidazolidone, 96%, anhydrous
2-Imidazolidone, 96%, anhydrous
2-Imidazolidone, 96%, anhydrous
Thermo Scientific Chemicals

2-Imidazolidone, 96%, anhydrous

CAS: 120-93-4 | C3H6N2O | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 412270010
Price (EUR)
306,00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
306,00
Each
Chemical Identifiers
CAS120-93-4
IUPAC Nameimidazolidin-2-one
Molecular FormulaC3H6N2O
InChI KeyYAMHXTCMCPHKLN-UHFFFAOYSA-N
SMILESO=C1NCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only