N-Isopropylacrylamide, 99%, pure, stabilized
N-Isopropylacrylamide, 99%, pure, stabilized
N-Isopropylacrylamide, 99%, pure, stabilized
Thermo Scientific Chemicals

N-Isopropylacrylamide, 99%, pure, stabilized

CAS: 2210-25-5 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Catalog number 412780250
Price (EUR)
86,65
線上優惠
96,50
Save 9,85 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
86,65
線上優惠
96,50
Save 9,85 (10%)
Each
Chemical Identifiers
CAS2210-25-5
IUPAC NameN-(propan-2-yl)prop-2-enamide
Molecular FormulaC6H11NO
InChI KeyQNILTEGFHQSKFF-UHFFFAOYSA-N
SMILESCC(C)NC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Melting point63°C to 67°C
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only