DL-alpha-Methoxyphenylacetic acid, 99%
DL-alpha-Methoxyphenylacetic acid, 99%
DL-alpha-Methoxyphenylacetic acid, 99%
Thermo Scientific Chemicals

DL-alpha-Methoxyphenylacetic acid, 99%

CAS: 7021-09-2 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 413870010
Price (EUR)
70,60
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
70,60
Each
Chemical Identifiers
CAS7021-09-2
IUPAC Name2-methoxy-2-phenylacetic acid
Molecular FormulaC9H10O3
InChI KeyDIWVBIXQCNRCFE-UHFFFAOYNA-N
SMILESCOC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
Appearance (Color)Off-white to light beige
Appearance (Form)Crystalline powder
Melting point68°C to 72°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only