Methyl pivaloylacetate, 98%
Methyl pivaloylacetate, 98%
Methyl pivaloylacetate, 98%
Thermo Scientific Chemicals

Methyl pivaloylacetate, 98%

CAS: 55107-14-7 | C8H14O3 | 158.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number 414190010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS55107-14-7
IUPAC Namemethyl 4,4-dimethyl-3-oxopentanoate
Molecular FormulaC8H14O3
InChI KeyXTXCFTMJPRXBBC-UHFFFAOYSA-N
SMILESCOC(=O)CC(=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only