Methyl hexanoate, 99%, analytical standard for GC
Methyl hexanoate, 99%, analytical standard for GC
Methyl hexanoate, 99%, analytical standard for GC
Thermo Scientific Chemicals

Methyl hexanoate, 99%, analytical standard for GC

CAS: 106-70-7 | C7H14O2 | 130.19 g/mol
Have Questions?
Catalog NumberQuantity
4144200505 g
Catalog number 414420050
Price (EUR)
99,70
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
99,70
Each
Specifications
Acidity1mg KOH/g max.
Assay Percent Range98.5% min. (GC)
CAS106-70-7
ChEBICHEBI:77322
Chemical Name or MaterialMethyl hexanoate
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only