alpha-Naphtholbenzein
alpha-Naphtholbenzein
alpha-Naphtholbenzein
Thermo Scientific Chemicals

alpha-Naphtholbenzein

Indicator | CAS: 145-50-6 | C27H18O2 | 374.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 415320250
Price (EUR)
118,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
118,00
Each
Chemical Identifiers
CAS145-50-6
IUPAC Name(4E)-4-[(4-hydroxynaphthalen-1-yl)(phenyl)methylidene]-1,4-dihydronaphthalen-1-one
Molecular FormulaC27H18O2
InChI KeyVDDWRTZCUJCDJM-SOYKGTTHSA-N
SMILESOC1=C2C=CC=CC2=C(C=C1)C(\C1=CC=CC=C1)=C1/C=CC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to red-brown to brown-black
Appearance (Color)to orange-brown
Appearance (Form)Powder
Infrared spectrumConforms
Molar extinction coefficient>=50000 (0.01 g/l methanol) (207-213 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only