Pyridine, 99+%, ACS reagent
Pyridine, 99+%, ACS reagent
Pyridine, 99+%, ACS reagent
Thermo Scientific Chemicals

Pyridine, 99+%, ACS reagent

CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 418540010
Price (EUR)
170,65
キャンペーン価格
189,00
Save 18,35 (10%)
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
170,65
キャンペーン価格
189,00
Save 18,35 (10%)
Each
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.1 % (K.F.)
Ammonia (NH3)=<20 ppm
Appearance (Color)Clear colorless
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only