4-(2-Pyridylazo)resorcinol, 97+%
4-(2-Pyridylazo)resorcinol, 97+%
4-(2-Pyridylazo)resorcinol, 97+%
Thermo Scientific Chemicals

4-(2-Pyridylazo)resorcinol, 97+%

CAS: 1141-59-9 | C11H9N3O2 | 215.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 418590050
Price (EUR)
82,90
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
82,90
Each
Chemical Identifiers
CAS1141-59-9
IUPAC Name(4Z)-3-hydroxy-4-[2-(pyridin-2-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Molecular FormulaC11H9N3O2
InChI KeyVLCAILLZPUINNF-LCYFTJDESA-N
SMILESOC1=CC(=O)C=C\C1=N\NC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO4>=97.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only