D(+)-Sucrose, 99+%, for biochemistry, DNAse, RNAse and protease free
D(+)-Sucrose, 99+%, for biochemistry, DNAse, RNAse and protease free
D(+)-Sucrose, 99+%, for biochemistry, DNAse, RNAse and protease free
Thermo Scientific Chemicals

D(+)-Sucrose, 99+%, for biochemistry, DNAse, RNAse and protease free

D(+)-Sucrose, CAS # 57-50-1, is a disaccharide used in a variety of biochemical applications such as protein purification. It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose. | CAS: 57-50-1 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
View all
Fiber Drum
Catalog number 419760250
Price (EUR)
1.414,00
Each
Quantity:
25 kg
Packaging:
Fiber Drum
Request bulk or custom format
Price (EUR)
1.414,00
Each
Chemical Identifiers
CAS57-50-1
IUPAC Name(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyCZMRCDWAGMRECN-PWPRYFECNA-N
SMILESOC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<5 ppm
Appearance (Form)Crystals
Appearance (Color)White
Infrared spectrumConforms
Loss on drying=<0.03 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• D(+)-Sucrose is a disaccharide carbohydrate
• It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose


Applications

• D(+)-Sucrose can be used for protein and nucleic acid purification
• Its fatty acid esters have laboratory research applications in transdermal systems
• It is also used for sucrose-gradient centrifugation, which may be employed for the selective isolation of Nocardia species from soil

RUO – Research Use Only

General References:

  1. Jain, A.; Shore, A. M.; Jonnalagadda, S. C.; Ramanujachary, K. V.; Mugweru, A. Conversion of fructose, glucose, and sucrose to 5-hydroxymethyl-2-furfural over mesoporous zirconium phosphate catalyst. Appl. Catal. A 2015, 489, 72-76.
  2. Chen, Y.; Zhang, H.; Li, A.; Zhuo, K. Conductivities for ZnSO4/MgSO4+ sucrose/trehalose + water systems at 298.15 K. Fluid Phase Equilib. 2015, 388, 78-83.