Tetrabutylammonium phosphate monobasic, 99%, HPLC grade
Tetrabutylammonium phosphate monobasic, 99%, HPLC grade
Tetrabutylammonium phosphate monobasic, 99%, HPLC grade
Thermo Scientific Chemicals

Tetrabutylammonium phosphate monobasic, 99%, HPLC grade

CAS: 5574-97-0 | C16H38NO4P | 339.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 420160250
Price (EUR)
693,65
Online Exclusive
770,00
Save 76,35 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
693,65
Online Exclusive
770,00
Save 76,35 (10%)
Each
Chemical Identifiers
CAS5574-97-0
IUPAC Nametetrabutylazanium dihydrogen phosphate
Molecular FormulaC16H38NO4P
InChI KeyARRNBPCNZJXHRJ-UHFFFAOYSA-M
SMILESOP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Powder
UV(0.1 % in water) 1 cm cell
UVat 200 nm A: =<0.2
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography, Phase transfer catalyst
RUO – Research Use Only