2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%
Thermo Scientific Chemicals

2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid, 99%

CAS: 2154-67-8 | C9H14NO3 | 184.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 420640050
Price (EUR)
478,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
478,00
Each
Chemical Identifiers
CAS2154-67-8
IUPAC Name1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxylate
Molecular FormulaC9H14NO3
InChI KeySADYUZSTFFUBBP-UHFFFAOYSA-M
SMILESCC1(C)C=C(C([O-])=O)C(C)(C)N1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only