p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
Thermo Scientific Chemicals

p-Toluenesulfonic acid monohydrate, ACS reagent

CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol
Have Questions?
Catalog NumberQuantityPackaging
421211000100 gPlastic bottle
Catalog number 421211000
Price (EUR)
32,65
Online Exclusive
35,90
Save 3,25 (9%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
32,65
Online Exclusive
35,90
Save 3,25 (9%)
Each
Chemical Identifiers
CAS6192-52-5
IUPAC Name4-methylbenzene-1-sulfonic acid hydrate
Molecular FormulaC7H10O4S
InChI KeyKJIFKLIQANRMOU-UHFFFAOYSA-N
SMILESO.CC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water9.5 to 11.5 %
Titration with NaOH>=98.5 %
Clarity of solution(c=20, water) Passes test
Heavy metals (as Pb)=<0.001 %
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only