Ammonium sulfate, 99+%, ACS reagent
Ammonium sulfate, 99+%, ACS reagent
Ammonium sulfate, 99+%, ACS reagent
Thermo Scientific Chemicals

Ammonium sulfate, 99+%, ACS reagent

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
5 kg
25 kg
Packaging:
View all
Fibre drum
Catalog number 423400251
Price (EUR)
652,00
Each
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Price (EUR)
652,00
Each
Specifications
Assay Percent Range99+%
CAS7783-20-2
ChEBICHEBI:62946
Chemical Name or MaterialAmmonium sulfate
ColorSilver to Gray
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only