Ammonium thiocyanate, 98%, ACS reagent
Ammonium thiocyanate, 98%, ACS reagent
Ammonium thiocyanate, 98%, ACS reagent
Ammonium thiocyanate, 98%, ACS reagent
Thermo Scientific Chemicals

Ammonium thiocyanate, 98%, ACS reagent

CAS: 1762-95-4 | CH4N2S | 76.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
3 kg
Packaging:
Plastic bottle
Catalog number 423410030
Price (EUR)
224,00
Each
Quantity:
3 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
224,00
Each
Chemical Identifiers
CAS1762-95-4
IUPAC Nameammonium cyanosulfanide
Molecular FormulaCH4N2S
InChI KeySOIFLUNRINLCBN-UHFFFAOYSA-N
SMILES[NH4+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=97.5 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only