Barium carbonate, 99+%, ACS reagent, Thermo Scientific Chemicals
Barium carbonate, 99+%, ACS reagent, Thermo Scientific Chemicals
Barium carbonate, 99+%, ACS reagent, Thermo Scientific Chemicals
Barium carbonate, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium carbonate, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
4234400252.5 kgPlastic bottle
Catalog number 423440025
Price (EUR)
168,00
Each
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
168,00
Each
Add to cart
Chemical Identifiers
CAS513-77-9
IUPAC Namebarium(2+) carbonate
Molecular FormulaCBaO3
InChI KeyAYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES[Ba++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration excess HCl99.0 to 101.0 %
Heavy metals (as Pb)=<10 ppm
Insoluble matter=<0.015 % (in dilute HCl)
Oxidizing substance (as NO3)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only