Copper(II) acetate monohydrate, ACS reagent
Copper(II) acetate monohydrate, ACS reagent
Copper(II) acetate monohydrate, ACS reagent
Thermo Scientific Chemicals

Copper(II) acetate monohydrate, ACS reagent

CAS: 6046-93-1 | C4H8CuO5 | 199.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 423591000
Price (EUR)
36,90
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
36,90
Each
Specifications
Boiling Point240.0°C
CAS6046-93-1
Chemical Name or MaterialCopper(II) acetate monohydrate
ColorBlue-Green or Green
Density1.88g/mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only