Diphenylamine, ACS reagent
Diphenylamine, ACS reagent
Diphenylamine, ACS reagent
Thermo Scientific Chemicals

Diphenylamine, ACS reagent

CAS: 122-39-4 | C12H11N | 169.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 423650050
Price (EUR)
47,80
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
47,80
Each
Chemical Identifiers
CAS122-39-4
IUPAC NameN-phenylaniline
Molecular FormulaC12H11N
InChI KeyDMBHHRLKUKUOEG-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals or flakes
Nitrate (NO3)Passes test
Sensitivity(to nitrate) Passes test
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only