Nitromethane, 96%, ACS reagent
Nitromethane, 96%, ACS reagent
Nitromethane, 96%, ACS reagent
Thermo Scientific Chemicals

Nitromethane, 96%, ACS reagent

CAS: 75-52-5 | CH3NO2 | 61.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 424015000
Price (EUR)
78,65
Online exclusive
87,00
Save 8,35 (10%)
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
78,65
Online exclusive
87,00
Save 8,35 (10%)
Each
Chemical Identifiers
CAS75-52-5
IUPAC Namenitromethane
Molecular FormulaCH3NO2
InChI KeyLYGJENNIWJXYER-UHFFFAOYSA-N
SMILESC[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Infrared spectrumConforms
GC>=95 %
Water=<0.05 % (Coulometric)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only