Potassium dichromate, 99+%, ACS reagent
Potassium dichromate, 99+%, ACS reagent
Potassium dichromate, 99+%, ACS reagent
Thermo Scientific Chemicals

Potassium dichromate, 99+%, ACS reagent

CAS: 7778-50-9 | Cr2K2O7 | 294.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 424115000
Price (EUR)
111,65
Online exclusive
124,00
Save 12,35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
111,65
Online exclusive
124,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS7778-50-9
IUPAC Namedipotassium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular FormulaCr2K2O7
InChI KeyKMUONIBRACKNSN-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodometric>=99.0 %
Insoluble matter=<0.005 %
Chloride (Cl)=<10 ppm
Calcium (Ca)=<30 ppm
Sodium (Na)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only