Sodium nitrate, 99+%, ACS reagent
Sodium nitrate, 99+%, ACS reagent
Sodium nitrate, 99+%, ACS reagent
Thermo Scientific Chemicals

Sodium nitrate, 99+%, ACS reagent

CAS: 7631-99-4 | NNaO3 | 84.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
3 kg
10 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 424345000
Price (EUR)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7631-99-4
IUPAC Namesodium nitrate
Molecular FormulaNNaO3
InChI KeyVWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES[Na+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Titration after Ion exchange>=99.0 %
Iodate (IO3)=<5 ppm
Iron (Fe)=<3 ppm
Calcium (Ca)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only