Sodium sulfate, 99+%, ACS reagent, anhydrous
Sodium sulfate, 99+%, ACS reagent, anhydrous
Sodium sulfate, 99+%, ACS reagent, anhydrous
Thermo Scientific Chemicals

Sodium sulfate, 99+%, ACS reagent, anhydrous

CAS: 7757-82-6 | Na2O4S | 142.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 424410025
Price (EUR)
88,65
線上優惠
98,60
Save 9,95 (10%)
Each
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
88,65
線上優惠
98,60
Save 9,95 (10%)
Each
Chemical Identifiers
CAS7757-82-6
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline or granular powder
Potassium (K)=<100 ppm
Titration after Ion exchange>=99.0 %
Heavy metals (as Pb)=<5 ppm (by ICP-OES)
pH5.2 to 9.2 (5 % solution at 25.0°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only