Sodium thiosulfate pentahydrate, 99.5+%, ACS reagent
Sodium thiosulfate pentahydrate, 99.5+%, ACS reagent
Sodium thiosulfate pentahydrate, 99.5+%, ACS reagent
Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99.5+%, ACS reagent

CAS: 10102-17-7 | Na2O3S2·5H2O | 248.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 424460010
Price (EUR)
51,10
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
51,10
Each
Chemical Identifiers
CAS10102-17-7
IUPAC Namedisodium pentahydrate sulfanidesulfonate
Molecular FormulaH10Na2O8S2
InChI KeyPODWXQQNRWNDGD-UHFFFAOYSA-L
SMILESO.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration Iodimetric99.5 to 101.0 %
pH6.0 to 8.4 (5 % solution at 25.0°C)
SO4 and SO3 (as SO4)=<0.1 %
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only