5-Sulfosalicylic acid dihydrate, ACS reagent
5-Sulfosalicylic acid dihydrate, ACS reagent
5-Sulfosalicylic acid dihydrate, ACS reagent
Thermo Scientific Chemicals

5-Sulfosalicylic acid dihydrate, ACS reagent

CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 424510025
Price (EUR)
399,00
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
399,00
Each
Chemical Identifiers
CAS5965-83-3
IUPAC Name2-hydroxy-5-sulfobenzoic acid; bis(methane)
Molecular FormulaC9H14O6S
InChI KeyNFYHZVWMQHQKRU-UHFFFAOYSA-N
SMILESC.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.1 %
Salicylic acid=<0.04 % (HOC6H4COOH)
Insoluble matter=<0.02 %
Sulfate (SO4)=<0.02 %
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only