Cyclopentanecarboxaldehyde, 97%, stabilized
Cyclopentanecarboxaldehyde, 97%, stabilized
Cyclopentanecarboxaldehyde, 97%, stabilized
Thermo Scientific Chemicals

Cyclopentanecarboxaldehyde, 97%, stabilized

CAS: 872-53-7 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 425910050
Price (EUR)
145,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
145,00
Each
Chemical Identifiers
CAS872-53-7
IUPAC Namecyclopentanecarbaldehyde
Molecular FormulaC6H10O
InChI KeyVELDYOPRLMJFIK-UHFFFAOYSA-N
SMILESO=CC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Stabilizer0.1 % w/w hydroquinone
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4430 to 1.4480 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only