3-Chloro-4-pyridinecarboxaldehyde, 97%
3-Chloro-4-pyridinecarboxaldehyde, 97%
3-Chloro-4-pyridinecarboxaldehyde, 97%
Thermo Scientific Chemicals

3-Chloro-4-pyridinecarboxaldehyde, 97%

CAS: 72990-37-5 | C6H4ClNO | 141.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 426550010
Price (EUR)
41,80
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
41,80
Each
Chemical Identifiers
CAS72990-37-5
IUPAC Name3-chloropyridine-4-carbaldehyde
Molecular FormulaC6H4ClNO
InChI KeyZVGDKOQPJCOCLI-UHFFFAOYSA-N
SMILESClC1=C(C=O)C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
GC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only