L-Methioninamide hydrochloride, 98%
L-Methioninamide hydrochloride, 98%
L-Methioninamide hydrochloride, 98%
L-Methioninamide hydrochloride, 98%
Thermo Scientific Chemicals

L-Methioninamide hydrochloride, 98%

CAS: 16120-92-6 | C5H13ClN2OS | 184.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 426710010
Price (EUR)
78,70
Each
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
78,70
Each
Add to cart
Chemical Identifiers
CAS16120-92-6
IUPAC Name2-amino-4-(methylsulfanyl)butanamide hydrochloride
Molecular FormulaC5H13ClN2OS
InChI KeyPWCBMJHINDTXGV-UHFFFAOYNA-N
SMILESCl.CSCCC(N)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration Argentometric97.5 to 102.5 %
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
NMR (1H-NMR)Consistent with structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only