2-Bromo-N,N-diethylethylamine hydrobromide, 98%
2-Bromo-N,N-diethylethylamine hydrobromide, 98%
2-Bromo-N,N-diethylethylamine hydrobromide, 98%
Thermo Scientific Chemicals

2-Bromo-N,N-diethylethylamine hydrobromide, 98%

CAS: 1069-72-3 | C6H15Br2N | 261.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 426860250
Price (EUR)
109,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
109,00
Each
Chemical Identifiers
CAS1069-72-3
IUPAC Namehydrogen (2-bromoethyl)diethylamine bromide
Molecular FormulaC6H15Br2N
InChI KeyHLMHCDKXKXBKQK-UHFFFAOYSA-N
SMILES[H+].[Br-].CCN(CC)CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Titration Argentometric>=97.5 %
Appearance (Form)Powder or solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only