5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate, 96%
5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate, 96%
5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate, 96%
Thermo Scientific Chemicals

5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate, 96%

CAS: 129946-88-9 | C14H8F6O3S2 | 402.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 427670050
Price (EUR)
480,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
480,00
Each
Chemical Identifiers
CAS129946-88-9
IUPAC Name8-(trifluoromethyl)-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaen-8-ium trifluoromethanesulfonate
Molecular FormulaC14H8F6O3S2
InChI KeyQXXHXTRTGZBOGD-UHFFFAOYSA-M
SMILES[O-]S(=O)(=O)C(F)(F)F.FC(F)(F)[S+]1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or solid
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
HPLC>=95 %
Appearance (Color)White to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only