9-Bromo-9-phenylfluorene, 97%
9-Bromo-9-phenylfluorene, 97%
9-Bromo-9-phenylfluorene, 97%
Thermo Scientific Chemicals

9-Bromo-9-phenylfluorene, 97%

CAS: 55135-66-5 | C19H13Br | 321.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 428610010
Price (EUR)
42,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
42,00
Each
Chemical Identifiers
CAS55135-66-5
IUPAC Name9-bromo-9-phenyl-9H-fluorene
Molecular FormulaC19H13Br
InChI KeyHYQXNCDBSALQLB-UHFFFAOYSA-N
SMILESBrC1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
HPLC>=96.0 %
Melting point99°C to 103°C
NMR (1H-NMR)Consistent with structure
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only